786-082
786-082EA
340
EUR
InStock
786-082
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), OneQuant™
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Ideal for enzyme-antibody crosslinking.
OneQuant™ Sulfo SMCC: Single use vials to minimize waste.