Genomic DNA isolation and purification kits, OmniPrep™
Fournisseur: G Bioscienes
786-136
786-136EA 532 EUR InStock
786-136
Genomic DNA isolation and purification kits, OmniPrep™
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The OmniPrep™ kit isolates high quality genomic DNA from many different species and tissue types including animal, plant, bacteria, yeast, fungi, whole blood, and cells in culture. DNA can be isolated from samples high in polysaccharides or other contaminants that are difficult to remove from the DNA preparations.
  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Adaptable for variety of samples, including plant, bacteria, fungi, mammalian tissues, cells and blood
  • Simple two tube method, 20 to 40 minutes
  • No toxic chemicals, no phenol, no hazardous waste
  • Suitable for ~150 preparations, depending on sample
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité