Protease Screening Kit
Fournisseur: G Bioscienes
786-137
786-137EA 666 EUR InStock
786-137
Protease Screening Kit
Tests Tests enzymatiques
The Protease Screening Kit provides a simple and quick method for testing samples for proteolysis. Simply incubate sample in the reagent provided and obtain results.
The kit uses dye-labelled protein conjugate as protease substrate, which allows nanogram level detection. The absorbance of dye-labelled peptide is measured at 574 nm for determination of protease activity.
The kit is sufficient for 50 assays in a microwell format.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité