Fraction-FOCUS™
Fournisseur: G Bioscienes
786-168
786-168EA 1260 EUR InStock
786-168
Fraction-FOCUS™
Protéines extraction Protein Extraction Kits
Fraction-FOCUS™ utilises proven technology to fractionate and concentrate all proteomes into multiple fractions; simplifying 2D maps and enhancing detection of low abundant proteins. Fraction-FOCUS™ is fully compatible with all downstream protein identification techniques.
There is no detectable loss of total protein during the Fraction-FOCUS™ procedure. At the end of the fractionation, cellular proteins are in one of many fractions.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité