Protein fractionation kit, nuclear and cytoplasmic extraction kit
Fournisseur: G Bioscienes
786-182
786-182EA 602 EUR InStock
786-182
Protein fractionation kit, nuclear and cytoplasmic extraction kit
Protéines extraction Protein Extraction Kits
The Nuclear and Cytoplasmic Extraction kit is for the enrichment of cytoplasmic and nuclear fractions from cultured cells and tissues. The kit generates a cleaner separation of cytoplasmic proteins from nuclear proteins and is ideal for expression and transport studies.
  • For generation of cytosolic and ultra pure nuclear fractions
  • Provided with nuclear protein solubilisation buffers
This kit is based on organic buffers and contains a proprietary combination of various salts and agents. The kit is provided with reagents for solubilisation of nuclear fraction.
The kit is designed for >50 preps, depending on the tissue used, where one prep is 20×10⁶ mammalian cells or 100 mg mammalian tissue.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité