Proteome kits, mammalian proteome, FOCUS™
Fournisseur: G Bioscienes
786-246
786-246EA 439 EUR InStock
786-246
Proteome kits, mammalian proteome, FOCUS™
Protéines extraction Protein Extraction Kits
Extracts and solubilises nearly all of the proteins from mammalian samples, including membrane as well as soluble proteins, by a strong chaotropic extraction buffer to solubilise even the most difficult proteins. Suitable for biological samples from animal tissues and adherent and suspension cells.
  • Single step extraction protocol
  • Supplied with a strong proprietary chaotropic extraction buffer
  • For 50×100 mg animal tissue or 50 μl wet cell pellet preparations
  • Suitable for sample preparation for 2D gel electrophoresis and other applications
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité