Glycoprotein stain
Fournisseur: G Bioscienes
786-254
786-254EA 704 EUR InStock
786-254
Glycoprotein stain
Colorants Colorants pour gels d'electrophorèse
The stain uses an enhanced Periodic Acid-Schiff (PAS) method for detection of glycoprotein sugars. The supplied agent first oxidises the cis-diol sugar groups to aldehydes. The aldehyde groups react with the sensitive glycoprotein stain solution forming Schiff bonds and producing strong magenta colour bands. RAPIDstain™ can be used after glycoprotein staining to detect non-glycosylated proteins and the use of the stain enhances glycoprotein staining. The glycoprotein stainis highly convenient as all the key reagents required for staining are supplied and a unique positive & negative control is included.
  • Specific glycosylated protein staining with optional non-glycosylated protein staining
  • Two stains in one kit
  • Nanogram level sensitivity
  • Stable periodate/Schiff staining
Suitable for the highly sensitive detection of glycoproteins following gel electrophoresis or protein transfer to nitrocellulose membranes.
Information de livraison: Supplied with RAPIDstain™, an enhanced Coomassie stain.
Attention: Store Glyco‐positive and negative control at -20 °C and other components at 4 °C (Shipped at ambient temperature)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité