Genomic DNA isolation from tissues, OmniPrep™ for Tissue kit
Fournisseur: G Bioscienes
786-395
786-395EA 471 EUR InStock
786-395
Genomic DNA isolation from tissues, OmniPrep™ for Tissue kit
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
OmniPrep™ for Tissue Kit is to specifically isolate high quality genomic DNA from tissue samples, including fresh, frozen, fixed or paraffin-embedded tissue. The kit isolates DNA from bodily fluids, including plasma, serum, amniotic fluid, semen and CSF. Also isolates high quality DNA from cultured cells, non-mammalian nucleated blood and gram-negative bacteria.
  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Simple two tube method, 20 to 40 minutes
  • No toxic chemicals, no phenol, no hazardous waste
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité