SAM Methyltransferase assay, SAMfluoro™
Fournisseur: G Bioscienes
786-431
786-431EA 1350 EUR InStock
786-431
SAM Methyltransferase assay, SAMfluoro™
Tests Tests enzymatiques
The SAMfluoro™ assay is a fluorescent, continuous enzyme coupled assay that can continuously monitor SAM-dependent methyltransferases without the use of radioactive labels or endpoint measurements.
  • Detection of protein methylation or screening methylation inhibitors
  • Continuous enzyme coupled assay for kinetic studies
  • Fluorescent, non-radioactive assay
  • Supplied with all reagents, including positive control
  • Adaptable for enzymes that generate S-adenosylhomocysteine or 5’-methylthioadenosine
Information de livraison: The kit is supplied with the reagents for 100 assays and AdoHcy as a positive control.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité