Protease inhibitor mix
Fournisseur: Cytiva
80-6501-23
80-6501-23EA 360 EUR InStock
80-6501-23
Protease inhibitor mix
Suppléments et additifs pour culture cellulaire
Sample preparation often requires the inhibition of protease activity. Protease inhibitor mix combines competitive and non competitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.
  • Protease inhibitor cocktail specifically developed for sample preparation in 2-D studies
  • No EDTA is used, which allows optimal nuclease activity for removing nucleic acids from samples
  • Contains a mixture of both competitive and noncompetitive protease inhibitors that inhibit serine, cysteine, and calpain proteases - optionally, EDTA may be added to inhibit metalloproteases
  • Optimised concentration for excellent inhibition of protease activities
  • Effectively inhibits over 95% of protease activity
Protease inhibitor mix combines competitive and non competitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité