DPA/Terbium set pour test de fusion membranaire
Fournisseur: Biotium
80104
80104.EA 131 EUR InStock
80104.
DPA/Terbium set pour test de fusion membranaire
DPA/Terbium set
The principle of DPA/Tb3 for vesicle fusion assay is based on the fact that contact of the chelator dipicolinic acid (DPA) with terbium (III) forms an instant Tb3 -DPA complex that is ~10,000 times more fluorescent than free Tb3 . In the assay, separate vesicle populations are loaded with 2.5 mM TbCl3 in 50 mM sodium citrate, or 50 mM DPA in 20 mM NaCl.
  • λEx/λEm (DPA/Tb3 complex) = 276/490 and 545 nm
  • DPA and Terbium are white solids and readily soluble in water
  • Store both reagents at room temperature
Fusion of the two types of vesicles results in fluorescence increase at 490 nm or 545 nm, with excitation at276 nm (1-4). Each set of product contains 1 g terbium trichloride and 1 g DPA in two separate vials. Please also see SDIP/Europium for membrane fusion assay (80105), which results in intense red fluorescence upon complex formation.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité