Colonnes SPE, J.T.Baker®, BAKERBOND Speedisk® C18 PolarPlus®
Fournisseur: Avantor
8153-04EA 329 EUR InStock
8153-04
Colonnes SPE, J.T.Baker®, BAKERBOND Speedisk® C18 PolarPlus®
Colonnes pour chromatographie
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Non ionic, basic, non polar to polar analytes.
These SPE columns can be used in reversed phase mode.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité