Packaging tape
Fournisseur: DESCO
242210
819-0142EA 9.1 EUR InStock
819-0142
Packaging tape
Rubans adhésifs
Desco Europe packaging tape is a conventional self-adhesive PVC tape.
  • Suitable for sealing secondary packaging such as cartons
  • Conventional PVC self adhesive yellow tape, not for use in an EPA
  • Printed black with logo and text to EN 61340-5
It uses an acrylic gum adhesive. The tape is yellow and is printed in black with the ESD Sensitivity Symbol ( Ref: EN 61340-5-1) alerting that package contents are ESD sensitive and should be opened only by a grounded operator at an ESD protective workstation.
The tape is tribocharging and is not suitable for use in an EPA. It is however suitable for sealing secondary packaging of electrostatic sensitive devices such as cardboard boxes.
Tape printed with wording: Attention Observe precautions for handling Electrostatic Sensitive Devices.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité