ESD high temp masking tape, paper core, Wescorp
Fournisseur: DESCO
242280
819-0148EA 6.8 EUR InStock
819-0148
ESD high temp masking tape, paper core, Wescorp
Rubans adhésifs
Economical crepe paper tape (7.0 mils thick) with high-tack rubber adhesive are used in applications masking PCB gold features for wave soldering.
  • Lead-free RoHS CompliantWaterproof
  • Non-toxic, non-corrosive
  • pH neutral
  • Tensile strength: 12 mm
  • Adhesion to steel: 155,04 g/cm²
  • Elongation: 8%
  • Temperature range: −36 to 135 °C (−32 to +275 °F)
  • Excellent conformability
Wescorp tape does tribocharge, but has minimal charge retention, making it safe to use around most ESD sensitive items. May be exposed to high temperatures for an extended period of time without leaving residue when removed.
Temperature resistance to a maximum of 135 °C (275 °F) for 45 min. Film resistance: <1×10¹¹ ohms tested per ANSI/ESD S11.11 at 50% RH.
Adhesive mass: natural rubber, non-staining.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité