84415-500ML
84415-500MLEA
83.8
EUR
InStock
84415-500ML
Acide chlorhydrique fumant ≥37% (par analyse titrimétrique), TraceSELECT™ pour analyses de traces, Fluka™
Acide chlorhydrique
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions