85850.270EA
111
EUR
InStock
85850.270
Acide nitrique fumant ≥99%, AnalaR NORMAPUR®
Acide nitrique
Packed in a plastic coated amber glass bottle with a vented cap.
Information de livraison: For transport the bottle is packed in a cardboard box, in a moulded fibre tray with Vermiculite, in a plastic bag, in a metal can.
Attention: According to the chemical properties of 100% nitric acid the influence of light can cause formation of nitrogen oxides.