86C-1KT
86C-1KTEA
341
EUR
InStock
86C-1KT
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-BI and fast blue BB salt, Sigma-Aldrich®
Leukocyte Alkaline Phosphatase Kit
Leukocyte Alkaline Phosphatase (LAP) kits are intended for the semi-quantitative demonstration of alkaline phosphatase activity in white blood cells.
- Suitable for haematology
- Suitable for histology
Peripheral blood or bone marrow preparations are fixed to a microscope slide. The film is then incubated in a mixture of naphthol AS-BI alkaline solution with fast blue BB. The resulting insoluble diffuse, blue dye deposit indicates sites of alkaline phosphatase activity.
Kit component
Citrate Solution (91-5) 50 mL
FBB-Alkaline Solution (86-3) 10 mL
Naphthol AS-BI Alkaline Solution (86-1) 10 mL
Neutral Red Solution (N6264) 50 mL
Sodium Nitrite Solution (91-4) 10 mL