87508
87516
87519
87500
87515
87504
87506
87511
87514
87520
87501
87505
87510
87507
87509
87503
87517
87518
87512
87502
87513
87508.EA
288
EUR
InStock
87508.
87516.
87519.
87500.
87515.
87504.
87506.
87511.
87514.
87520.
87501.
87505.
87510.
87507.
87509.
87503.
87517.
87518.
87512.
87502.
87513.
Easy Dry - Petri dish with dehydrated medium in pads
Milieux pour microbiologie
Milieux pour microbiologie déshydratés
Easy Dry are absorbent pads impregnated with a sterile, dehydrated culture medium.
- Available aslo with chromogenic culture media for an easy differentiation of microorganisms
- Ready to use; just add sterile water
- Storage at 10 to 25 °C
- Shelf life: 2 years from the production date
Easy Dry pads are preplated in a Petri dish and is immediately ready to use after pouring sterile distilled or deionised water on it. Easy Dry are optimal for the examination of large sample volumes by the membrane filter method.