Easy Dry - Petri dish with dehydrated medium in pads
Fournisseur: LIOFILCHEM S.R.L.
87508 87516 87519 87500 87515 87504 87506 87511 87514 87520 87501 87505 87510 87507 87509 87503 87517 87518 87512 87502 87513
87508.EA 288 EUR InStock
87508. 87516. 87519. 87500. 87515. 87504. 87506. 87511. 87514. 87520. 87501. 87505. 87510. 87507. 87509. 87503. 87517. 87518. 87512. 87502. 87513.
Easy Dry - Petri dish with dehydrated medium in pads
Milieux pour microbiologie Milieux pour microbiologie déshydratés
Easy Dry are absorbent pads impregnated with a sterile, dehydrated culture medium.
  • Available aslo with chromogenic culture media for an easy differentiation of microorganisms
  • Ready to use; just add sterile water
  • Storage at 10 to 25 °C
  • Shelf life: 2 years from the production date
Easy Dry pads are preplated in a Petri dish and is immediately ready to use after pouring sterile distilled or deionised water on it. Easy Dry are optimal for the examination of large sample volumes by the membrane filter method.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité