Synonyms:
Sulphuric acid-[²H₂]
87857.18
87857.18EA
242
EUR
InStock
87857.18
Sulphuric acid-[D₂] 96% (99.5% D) (p/p) pour spectroscopie RMN
Sulphuric acid-[D₂]
Purity: 96% min
CAS No.: 13813-19-9
Molecular Formula: D2SO4
Formula Weight: 100.09
Physical Form: Liquid
Density: 1.86
Refractive Index: 1.4175
Sensitivity: Moisture Sensitive
Odour: Odourless
Boiling Point: 290°
UN No.: 1830
MDL No.: MFCD00003542