87862.290EA
299
EUR
InStock
87862.290
Magnésium nitrate solution solution Reag. Ph. Eur. 1049801
Magnesium nitrate solutions
The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water.
Starting materials: magnesium nitrate R / water R / ethanol (96 per cent)
Informations de commande: Livré avec un certificat d'analyse. Fabriqués sur commande. Verifiez le stock avec votre service client.