88380.290EA
918
EUR
InStock
88380.290
Bore 5,000 mg/l solution étalon dans l'eau
Étalons
Étalons cationiques
Standards de bore
This certified reference material is produced using a high-purity starting material, acid from sub-boiling and 18 MOhm deionised water.