88458.18
88458.18EA
1290
EUR
InStock
88458.18
Arséniure de gallium ≥99.999% (base métaux)
Arséniure de gallium
Purity: 99.999%
CAS No.: 1303-00-0
Molecular Formula: GaAs
Formula Weight: 144.64
Physical Form: 25mm & Down Polycrystalline Pieces
Density: 5.31
Melting Point: 1238°
Merck Reference: 14,4347
UN No.: 1557
MDL No.: MFCD00011017
Semiconductors (transistors, lasers, solar cells)