Synonyms:
Aluminum iodide
89185.06
89185.06EA
148
EUR
InStock
89185.06
Iodure d'aluminium ≥99.99% (base métaux)
Iodure d'aluminium
Purity: 99.99+%
CAS No.: 7784-23-8
Molecular Formula: AlI3
Formula Weight: 407.69
Physical Form: 2-8mm Lump
Density: 3.98
Appearance: White to brownish-purple
Odour: Pungent
Melting Point: 191°
Boiling Point: 360°
Merck Reference: 14,346
UN No.: 3260
MDL No.: MFCD00003427