Supelco® Organic Reference Standard, BDE No 209 solution
Fournisseur: Merck
89407-1ML
89407-1MLEA 181 EUR InStock
89407-1ML
Supelco® Organic Reference Standard, BDE No 209 solution
Étalons Étalons organiques
The customer needs to make sure that the standard is homogeneous this can be achieved by shaking vortexing or sonicating the ampoule before opening the ampoule.
Care should be taken when opening the ampoule to avoid any injuries from the broken glass. The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent. After opening the ampoule the standard should not be stored or kept in the ampoule.
To preserve the integrity of the product the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.
Attention: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité