Synonyms:
Methylbenzene, Phenylmethane, FM approval
89681-10L
89681-10LEA
419
EUR
InStock
89681-10L
Toluène ≥99.7% (par GC), Puriss. p.a. ACS, Riedel-de Haën™
Toluène
ACS GENERAL USE SOLVENTS
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested to meet ACS general use specifications
Beilstein Number: 635760