Reactive dye conjugates
Fournisseur: Biotium
90073 92000 90103 90104 90105 90106 90107 90108 90109 90110 90115 90116 92001
90073.EA 368 EUR InStock
90073. 92000. 90103. 90104. 90105. 90106. 90107. 90108. 90109. 90110. 90115. 90116. 92001.
Reactive dye conjugates
Systèmes de liaison et de marquage des protéines
Reactive dyes containing an amine functional group.
ARP (N-(Aminooxyacetyl)-N’-(D-biotinoyl)hydrazine, trifluoroacetic acid salt) reacts with the exposed aldehyde group formed at abasic sites in damaged DNA, allowing the DNA to be labeled with biotin groups. The labeled DNA can then be quantitated with fluorescent or enzyme-conjugated streptavidin complexes. ARP can freely cross the cell membranes, thus allowing detection of abasic sites in living cells. ARP is suitable for use in microplate assays.
Aminooxy reagents, such as aminooxy-5(6)-FAM, react with aldehydes or ketones to form a stable hydrazone compound under mild conditions. Similar to ARP, aminooxy-5(6)-FAM can be used to label abasic sites in damaged DNA.
Dye conjugates to PEO3-amine have a long water-soluble PEG spacer that separates the fluorescent dye and the amine functional group. These dyes can be coupled to activated carboxy groups or sulfonyl chlorides. Cadaverine conjugates can be coupled to activated carboxy groups or sulfonyl chlorides.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d407289e04a69b6db73564694b104e91b034123b&vid=d6e1ed068d1d70ae48acaf9e956e4ece449619a1&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1670a8bfeee9933a538ab1f4e21d4ce43b55614&vid=8c7ea04645033ef7cb5562295a72b9360fa762a3&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656f4e29aef745b7242e9d25ff604a71a8c51bcb&vid=9985e76b5efb11aa38b212773b6773f4b887bf1f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec31c4fb5558a73afabbad4658af46d0493c5019&vid=b0e61ed2ca72a01a18c15bc1a671886d66018070&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a5ada650030d1a3ddfcae3ca527941cd829d67d&vid=df189fa4871932f49c9669f7483c7a4c5090075d&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3831e02d51a046a7b2eab93f038ccb1e8a1e21c6&vid=365f1e1053dee4fed5480c29ccf2dcd682749cbf&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a90d89bc32ba6538a733645994582aeaf0a2a980&vid=5390cc3f7a77c0af18f531f702213db7ede9f5fb&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e27bd253acb119e7be58a64956d9acbb64b85931&vid=62835b47782a94c30844d6994c84455edd0616f1&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06f4e03ac8002fbcb096f4960ef1726407b5a8cd&vid=3ecc3a25a41c6c5f396aba822f17c475e94dbd07&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e2ec0de2c1e9a99c4a0f765899ead91809e395b&vid=e4115b3f603bf41086b08034e5cad5c9bcba0af5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81ca38bf6c75d608a476a16ef82e7a81deaf000b&vid=c2b225ce55749628fa9ad2bf55007873c36f6293&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fc50a26092a1e91234bfabe611f26b91273baa8&vid=33cc22ceeded205d3b2fc102ebb90ecb24568ec4&prid=web&clid=SAPDAM
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité