6-((7-Amino-4-methylcoumarin-3-acetyl) amino)hexanoic acid succinimidyl ester (AMCA-X SE) colorant fluorescent réactif aux amines
Fournisseur: Biotium
90077
90077.EA 268 EUR InStock
90077.
6-((7-Amino-4-methylcoumarin-3-acetyl) amino)hexanoic acid succinimidyl ester (AMCA-X SE) colorant fluorescent réactif aux amines
6-((7-Amino-4-methylcoumarin-3-acetyl) amino)hexanoic acid succinimidyl ester (AMCA-X SE)
AMCA-X SE is an amine-reactive, UV-excitable, blue fluorescent dye.
  • λEx/λ Em(MeOH) = 353/442 nm
  • White solid soluble in DMSO or DMF
  • Store at -20°C and protect from light
AMCA-X SE is an amine-reactive blue fluorescent dye widely used for labeling proteins or other biomolecules that contain a primary or secondary aliphatic amine. A stock solution of the dye can be made in anhydrous DMF or DMSO. The coupling reaction is usually carried out in a pH ~8.5 buffer (for example, 0.1-0.2 M sodium bicarbonate buffer). Alternatively, the coupling reaction may also be carried out in an organic solvent for amino compounds that are insoluble in aqueous solution.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité