Supelco® TraceCERT® Organic Reference Standard, Imazamethabenz-methyl
Fournisseur: Merck
90556-50MG
90556-50MGEA 95.3 EUR InStock
90556-50MG
Supelco® TraceCERT® Organic Reference Standard, Imazamethabenz-methyl
Étalons Étalons organiques
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The imazamethabenz-methyl CRM may also find following uses:
  • Study the photolysis of imazamox imazapic imazaquin and imazamethabenz-methyl as functions of wavelength pH and natural organic matter in an aqueous solution
  • Comparative study of the extraction of imazamethabenz-methyl from agricultural soils followed by its chromatographic determination using HPLC
  • Investigate the absorption and translocation of fenoxaprop-ethyl and imazamethabenz-methyl in wild oat plants grown under different conditions of temperature and light intensity using ¹⁴C tracer techniques
  • Simultaneous determination of p- and m-imazamethabenz-methyl enantiomers using a protein chiral selector based on a direct chiral HPLC method
  • To develop a multiresidue method for the determination of post-emergence herbicides in water samples by SPE coupled with HPLC/ESI/MS in positive ionisation mode
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité