Supelco® TraceCERT® Organic Reference Standard, Iprobenfos
Fournisseur: Merck
91386-100MG
91386-100MGEA 208 EUR InStock
91386-100MG
Supelco® TraceCERT® Organic Reference Standard, Iprobenfos
Étalons Étalons organiques
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The iprobenfos CRM may also find following uses:
  • Sensitive detection of iprobenfos and edifenphos using gold nanoparticle AuNP-based colorimetric multi-aptasensor from rice samples
  • To investigate light-induced catalytic degradation of iprobenfos fungicide using poly(3-octylthiophene-25-diyl) film by varying oxygen pressure of bubbling gases
  • To determine the variation in sensitivity of Magnaporthe oryzae isolates from different geographic areas towards edifenphos and iprobenfos fungicides
  • To study the response of zooplankton community against the application of simetryn and iprobenfos using natural zooplankton community in microcosm tanks
  • To evaluate and compare the virulence of Magnaporthe oryzae isolates sensitive and resistant to edifenphos and iprobenfos on rice plants treated with or without fungicides
Attention: P264 - P270 - P273 - P301 + P312 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité