Supelco® TraceCERT® Organic Reference Standard, Isazophos
Fournisseur: Merck
91607-100MG
91607-100MGEA 125 EUR InStock
91607-100MG
Supelco® TraceCERT® Organic Reference Standard, Isazophos
Étalons Étalons organiques
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isazophos CRM may also find its use as described below:
  • To develop a GC/MS procedure utilising selected ion monitoring (SIM) for quantification of isazophos in bovine liver kidney muscle and fat at various times after exposure
  • To develop multiresidue method for quantification of 19 metabolites of organophosphorus pesticides synthetic pyrethroids selected herbicides and DEET (NN-diethyl-m-toluamide) in human urine using LC-MS
  • To determine the efficacy of isazophos against Japanese beetle (Popillia japonica Newman) grubs using two subsurface placement technology high-pressure liquid injection (HPLI) and an adapted overseeder (OS) applied to golf course roughs
Attention: P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310 - P314
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité