Phytochemical standard, Nobiletin, analytical standard, Supelco®
Fournisseur: Merck
92600-10MG
92600-10MGEA 606 EUR InStock
92600-10MG
Phytochemical standard, Nobiletin, analytical standard, Supelco®
Étalons Étalons organiques
Nobiletin suppresses the expression of matrix metalloproteinases 1, 3 and 9, which participate in the break down of the extracellular matrix during tumor metastasis. Nobiletin is also involved in the increased expression of tissue inhibitor of matrix metalloproteinase 1 further preventing extracellular matrix degradation during tumor invasion. This bioflavonoid is believed to achieve both of these functions by disrupting the signal transduction pathway of Ras-Raf-MEK.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fededd68ea4808d99fa3f571022795beab0822ab&vid=fededd68ea4808d99fa3f571022795beab0822ab&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité