Organic reference standard, Zinc oxide, analytical standard, Supelco®
Fournisseur: Merck
93632-100MG
93632-100MGEA 82.6 EUR InStock
93632-100MG
Organic reference standard, Zinc oxide, analytical standard, Supelco®
Étalons Étalons organiques
Zinc oxide has been used as a raw material in the synthesis of environmental friendly silver front contact paste comprising of silver particles, glass frit powders, and resin binder, which can be used in crystalline silicon solar cells.
Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.
Conditionnement: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453e0533a0458899836428b5c43e83e946dac6c6&vid=453e0533a0458899836428b5c43e83e946dac6c6&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité