Synonyms:
Chloranilicacid
A10493.14
A10493.14EA
44.1
EUR
InStock
A10493.14
2,5-Dichloro-3,6-dihydroxybenzoquinone ≥98%
2,5-Dichloro-3,6-dihydroxybenzoquinone
Purity: 98+%
CAS No.: 87-88-7
Molecular Formula: C6H2Cl2O4
Formula Weight: 208.99
Melting Point: 300° dec.
Beilstein Reference: 1875040
Merck Reference: 14,2079
MDL No.: MFCD00001596