Synonyms:
2-(Trifluoromethyl)benzoyl chloride, 2-(Trifluoromethyl)benzene-1-carbonyl chloride
A10564.A1
A10564.A1EA
0
EUR
InStock
A10564.A1
Chlorure d'α,α,α-trifluoro-o-toluoyle ≥98%
Chlorure d'α,α,α-trifluoro-o-toluoyle
Purity: 98%
CAS No.: 312-94-7
Molecular Formula: C8H4ClF3O
Formula Weight: 208.57
Flash Point: 95°(203°F)
Refractive Index: 1.4790
Odour: Pungent
Melting Point: -22°
Boiling Point: 89-90°/16mm
Beilstein Reference: 881040
UN No.: 3265
MDL No.: MFCD00000667