Synonyms:
2-Cyano-N,N-dimethylacetamide
A10981.06
A10981.06EA
58.2
EUR
InStock
A10981.06
N,N-Dimethylcyanoacetamide ≥98%
N,N-Dimethylcyanoacetamide
Purity: 98%
CAS No.: 7391-40-4
Molecular Formula: NCCH2CON(CH3)2
Formula Weight: 112.13
Melting Point: 59-66°
Beilstein Reference: 773950