Synonyms:
2-Fluoro-3-methylbenzoic acid
A12280.03
A12280.06
A12280.03EA
45.8
EUR
InStock
A12280.03
A12280.06
2-Fluoro-m-toluic acid ≥98%
2-Fluoro-m-toluic acid
Purity: 98%
CAS No.: 315-31-1
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 114-116°
MDL No.: MFCD00042504