A13243.22
A13243.22EA
49
EUR
InStock
A13243.22
Chlorure de piperidinium ≥99%
Chlorure de piperidinium
Purity: 99%
CAS No.: 6091-44-7
Molecular Formula: C5H11N.HCl
Formula Weight: 121.61
Flash Point: 110°(230°F)
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 245-248°
Beilstein Reference: 3611699
UN No.: 2811
MDL No.: MFCD00012770