A13281.30
A13281.30EA
114
EUR
InStock
A13281.30
Acide 2-benzoylbenzoïque ≥98%
Acide 2-benzoylbenzoïque
Purity: 98+%
CAS No.: 85-52-9
Molecular Formula: C14H10O3
Formula Weight: 226.23
Odour: Odourless
Melting Point: 127-130°
Boiling Point: 257°
Beilstein Reference: 1107841
MDL No.: MFCD00002472