Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A13397.AP
A13397.APEA
59
EUR
InStock
A13397.AP
Ether diméthylique du diéthylèneglycol ≥99% stabilisé
Ether diméthylique du diéthylèneglycol
Stabilized with 100 ppm BHT
Purity: 99%
CAS No.: 111-96-6
Molecular Formula: C6H14O3
Formula Weight: 134.18
Flash Point: 51°(124°F)
Refractive Index: 1.4080
Odour: Ether-like
Melting Point: -64°
Boiling Point: 161-162°
Beilstein Reference: 1736101
Merck Reference: 14,3165
UN No.: 3271
MDL No.: MFCD00008503