A13556.22
A13556.22EA
353
EUR
InStock
A13556.22
1,1,1-Trifluoroacétone 95%
1,1,1-Trifluoroacétone
Purity: 95%
CAS No.: 421-50-1
Molecular Formula: CF3COCH3
Formula Weight: 112.05
Storage Temperature: 2°C to 8°C
Flash Point: -31°(-24°F)
Refractive Index: 1.3000
Odour: Chloroform-like
Melting Point: <-78°
Boiling Point: 21-24°
Beilstein Reference: 1748614
UN No.: 1993
MDL No.: MFCD00000423