A14089.30
A14089.0B
A14089.0I
A14089.30EA
59.2
EUR
InStock
A14089.30
A14089.0B
A14089.0I
Triphénylphosphine 99%, en paillettes
Triphénylphosphine
Purity: 99%
CAS No.: 603-35-0
Molecular Formula: (C6H5)3P
Formula Weight: 262.29
Flash Point: 181°(358°F)
Physical Form: Flakes
Density: 1.200
Appearance: White to pale cream
Odour: Mild
Melting Point: 79-81°
Boiling Point: 360°
Solubility: Insoluble in water. Soluble in benzene, acetone, carbon tetrachloride, ether, and glacial acetic acid
Beilstein Reference: 610776
Merck Reference: 14,9743
MDL No.: MFCD00003043
In the synthesis of organic compounds, phosphonium salts and other phosphorus compounds, and as a polymerization initiator