Synonyms:
3,4-Dichlorobenzene-1-sulphonyl chloride
A14576.06
A14576.06EA
65.9
EUR
InStock
A14576.06
Chlorure de 3,4-dichlorobenzènesulfonyle ≥98%
Chlorure de 3,4-dichlorobenzènesulfonyle
Purity: 98%
CAS No.: 98-31-7
Molecular Formula: C6H3Cl3O2S
Formula Weight: 245.51
Flash Point: 110°(230°F)
Refractive Index: 1.5890
Melting Point: 20-22°
Boiling Point: 253°
Beilstein Reference: 1956417
UN No.: 3265
MDL No.: MFCD00041255