Synonyms:
2-(4-Chlorophenoxy)acetonitrile
A15389.14
A15389.14EA
76.2
EUR
InStock
A15389.14
(4-Chlorophenoxy)acetonitrile ≥98%
(4-Chlorophenoxy)acetonitrile
Purity: 98%
CAS No.: 3598-13-8
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Melting Point: 45-49°
Boiling Point: 149-150°/15mm
UN No.: 3439
MDL No.: MFCD00031535