A15997.30
A15997.18
A15997.0B
A15997.30EA
82.8
EUR
InStock
A15997.30
A15997.18
A15997.0B
Theophyllin monohydrate ≥99%
Theophyllin monohydrate
Purity: 99%
CAS No.: 5967-84-0
Molecular Formula: C7H8N4O2.H2O
Formula Weight: 198.18 (180.16anhy)
Odour: Odourless
Melting Point: 270-274°
Beilstein Reference: 13463
Merck Reference: 14,9285
UN No.: 2811
MDL No.: MFCD00151659