A16029.14
A16029.14EA
454
EUR
InStock
A16029.14
4-Chloro-3-fluorophénol ≥98%
4-Chloro-3-fluorophénol
Purity: 98%
CAS No.: 348-60-7
Molecular Formula: C6H4ClFO
Formula Weight: 146.55
Flash Point: >110°(230°F)
Odour: Phenol-like
Boiling Point: 84°/44mm
Beilstein Reference: 3236440
UN No.: 3261
MDL No.: MFCD00042583