A16214.0E
A16214.0EEA
419
EUR
InStock
A16214.0E
Fumarate de diéthyle ≥98%
Fumarate de diéthyle
Purity: 98%
CAS No.: 623-91-6
Molecular Formula: C8H12O4
Formula Weight: 172.18
Flash Point: 91°(195°F)
Refractive Index: 1.4400
Melting Point: ca 0°
Boiling Point: 98-100°/14mm
Beilstein Reference: 775347
MDL No.: MFCD00064455