Synonyms:
1,2-Dibromo-3,4,5,6-tetrafluorobenzene
A16620.03
A16620.03EA
62.8
EUR
InStock
A16620.03
1,2-Dibromotétrafluorobenzène ≥99%
1,2-Dibromotétrafluorobenzène
Purity: 99%
CAS No.: 827-08-7
Molecular Formula: C6Br2F4
Formula Weight: 307.87
Flash Point: 99°(210°F)
Refractive Index: 1.5160
Boiling Point: 197-199°
MDL No.: MFCD00039209