Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 250 ppm
A16833.14
A16833.14EA
212
EUR
InStock
A16833.14
2,3,4,5,6-Pentafluorostyrène ≥98% stabilisé
2,3,4,5,6-Pentafluorostyrène
Stabilized with 250 ppm 4-tert-butylcatechol
Purity: 98%
CAS No.: 653-34-9
Molecular Formula: C8H3F5
Formula Weight: 194.10
Storage Temperature: 2°C to 8°C
Flash Point: 34°(93°F)
Refractive Index: 1.4460
Odour: Characteristic
Boiling Point: 139-140°
Beilstein Reference: 1874390
UN No.: 1993
MDL No.: MFCD00000300