A17416.36
A17416.36EA
28.8
EUR
InStock
A17416.36
di-Ammonium hydrogénoorthophosphate ≥98%
di-Ammonium hydrogen orthophosphate
Purity: 98%
CAS No.: 7783-28-0
Molecular Formula: H9N2O4P
Formula Weight: 132.06
Melting Point: 155° dec.
Merck Reference: 14,542
MDL No.: MFCD00010891