Synonyms:
Tetramethylammoniumhexafluorophosphate
A17467.22
A17467.22EA
334
EUR
InStock
A17467.22
Hexafluorophosphate de tétraméthylammonium 99%
Hexafluorophosphate de tétraméthylammonium
Purity: 99+%
CAS No.: 558-32-7
Molecular Formula: C4H12F6NP
Formula Weight: 219.10
Odour: Odourless
Melting Point: 300°
Beilstein Reference: 3730394
UN No.: 2923
MDL No.: MFCD00012012